Seismograph Model
Supplier: Wards
BG0014
470324-782EA 64.25 USD
470324-782
Seismograph Model
Models Geology Models
Clearly demonstrate how a seismograph records the shock waves generated by an earthquake in the classroom or laboratory.
  • Heavy duty, premium seismograph model
  • Can be used to study both geological and mathematical phenomena
  • Writing instrument is not included
This seismograph model creates a visual representation of the shock waves generated by an earthquake. It can be used to teach both mathematical and geological concepts in an educational setting. Demonstration is conducted by manually pulling the paper strip and recording simple movements made by jumping or moving next to the model. This model is designed for educational concept demonstration and is not intended to be used to accurately measure seismic waves.