Celestial Star Globe
Supplier: Wards
BD0077CSG
470324-780EA 93.35 USD
470324-780
Celestial Star Globe
Globes
Perfect for introductory-level astronomy students.
  • 12" Globe diameter
  • 17.5" Tall (overall model)
  • A 3" detailed earth globe is fitted inside the large star globe
This stunning star globe illustrates the effects of Earth’s rotation on its axis and its revolution around the sun in addition to depicting the constellations, Milky Way, and major stars in three dimensions. Color-coding and clear silk-screened names make it easy to identify the many stars, while celestial latitude and longitude scales provide orientation. A detailed 3" Earth globe is centered in the larger star globe. Both globes can be set to the positions of the stars and planets for any time and place on Earth in the past, present, or future.