HOBO® DC Voltage Input Cable, 0 to 5 V
Supplier: ONSET COMPUTER CORPORATION SE
CABLE-ADAP5
470324-392EA 66.99 USD
470324-392
HOBO® DC Voltage Input Cable, 0 to 5 V
Cables
Connects 3rd-party sensors with a DC Voltage ouput to external-channel data loggers.
  • Expands the range of measurement options and applications
Cable length is 1.8 m (6') cable with 13 mm (0.5") tinned braided wire leads.
Ordering information: Connects 3rd-party sensors with a DC Voltage ouput to a HOBO 12, UX120, MX1100, or ZW series external-channel data logger.