mBot Ultimate 2.0 Robot Kit
Supplier: THE DOUGLAS STEWART CO.,INC
121868
470324-128EA 429.99 USD
470324-128
mBot Ultimate 2.0 Robot Kit
Educational Materials Physics Educational Materials Applied Physics Learning Activities
This 10-in-1 robot kit offers exciting functionality and compatibility.
  • Chassis: Aluminum alloy
  • Processor: ATMEGA2560-16AU
  • Coding language: Block-based, Arduino, Python (Extra Raspberry Pi Needed)
  • Communication mode: USB, Bluetooth, 2.4G
As an advanced programmable robotic kit, the mBot Ultimate 2.0 has more than 160 mechanical parts and modules, allows you to build 10 different configurations. Compatible with both Arduino and Raspberry Pi, the mBot Ultimate 2.0 is a DIY platform that can help you achieve practical and interesting projects.
Help students with engineering, mechanical, and programming learning.
Supports visual programming such as Scratch block-based programming, the Arduino IDE, and Python (Raspberry Pi available separately).