Turbitidy Tube
Supplier: GSC International
TURBTUB
470323-888EA 46.7 USD
470323-888
Turbitidy Tube
Turbidity Meters
Easily measure the cloudiness of a fluid.
  • Simple and accurately measures the turbidity of a fluid
To measure turbidity using this apparatus, fill the tube with water and drain it slowly by limiting flow with the pinch clamp on the hose. Stop draining once the black and white Secchi disk on the bottom of the tube becomes visible, and then take note of the water level using the graduations on the tube. Instructions included.