HOBOnet Rainfall Sensor (Metric)
Supplier: ONSET COMPUTER CORPORATION SE
RXW-RGF-900
470323-650EA 559.15 USD
470323-650
HOBOnet Rainfall Sensor (Metric)
Weather Stations
A wireless sensor that records rainfall in 0.2 millimeter increments.
  • 16 MB memory
  • Operating temperature range: –25 to 60 °C (–13 to 140 °F)
  • Powered by rechargeable AA batteries and a built-in solar pannel
This wireless sensor directly communicates data.
Ordering information: Only functional with specific HOBO systems. Contact the Science Help Desk for additional information (sciencehelp@avantorsciences.com).