Optic USB Base Station
Supplier: ONSET COMPUTER CORPORATION SE
BASE-U-4
470323-564EA 197.39 USD
470323-564
Optic USB Base Station
Data Loggers
The Universal Optic USB Base Station and couplers are used to offload data from any HOBO® data logger with an Optic USB interface.
  • Offload data from any HOBO® data logger with an Optic USB interface
  • Couplers are included
  • Cable length 183 cm (6')
  • Offloads full 64K logger in 30 seconds
Delivery information: Couplers for all compatible data loggers are included with the BASE-U-4. Couplers are included for compatibility with: UA Pendants, U20 water level data loggers, the U22 Water Temp Pro v2, our UTBI TidbiT, U24 Conductivit,y and U23 data loggers.