HOBO® State Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
UX90-001
470323-540EA 199.99 USD
470323-540
HOBO® State Data Logger
Data Loggers
Records state changes, electronic pulses, and mechanical or electrical contact closures from external sensing devices.
  • One logger with four recording modes
  • Internal reed switch can be used to determine when a door or window is open or closed
  • Compatible with a wide range of external sensors
  • Ships with 2.5 mm input cable
  • Compatible with HOBOware and HOBOware Pro software for logger setup and graphing/analysis
Ideal for monitoring energy consumption, mechanical equipment operation, and water and gas flow. 1 Hz maximum state, event, runtime frequency.