HOBO® 4-Channel Analog Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
UX120-006M
470323-530CT 73.75 USD
470323-530
HOBO® 4-Channel Analog Data Logger
Data Loggers
This logger supports up to four external sensors to record parameters like temp, current, etc.
  • Twice the accuracy over previous models with 16-bit resolution
  • Flexible support for a wide range of external sensors
  • LCD confirms logger operation and displays near real-time measurement data
  • Provides minimum, maximum, average, and standard deviation logging options
  • On-screen alarms notify you when a sensor reading exceeds set thresholds
  • Stores 1.9 million measurements for longer deployments between offloads
  • Compatible with HOBOware® and HOBOware® Pro software for logger setup, graphing, and analysis
A high-performance, LCD display data logger, supporting up to four external sensors to measure and record parameters such as temperature, current, CO₂, and voltage, for building performance monitoring applications. This logger is ideal for energy audits, building commissioning studies, equipment scheduling, and more.
Ordering information: Requires HOBOware® software and a USB cable. (Note: A USB cable is included with HOBOware® Pro CD or USB software purchase). See compatible items below.