HOBO® U23 Pro v2 External Temperature Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
U23-004
470323-516EA 216.79 USD
470323-516
HOBO® U23 Pro v2 External Temperature Data Logger
Data Loggers
A weatherproof data logger with an external soil/water temperature sensor on a 6' cable, for applications where just one temperature probe is required. Also includes an internal temperature sensor.
  • Weatherproof housing for use in outdoor or condensing environments
  • High accuracy
  • Small-diameter external sensor versions for measurements in tight spaces
  • Optic USB interface for fast and reliable download
Compatible with HOBOware® and HOBOware® Pro software for logger setup, graphing, and analysis.
Ordering information: Requires HOBOware® software and a USB cable. (NOTE: A USB cable is included with HOBOware® Pro CD or USB software purchase). HOBOware® Pro is required when using the U-DTW-1 waterproof shuttle.