HOBO® External Temperature Sensor Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
MX2304
470323-476EA 192.06 USD
470323-476
HOBO® External Temperature Sensor Data Logger
Data Loggers
The HOBO MX2304 features one 2 m external temperature sensor.
  • Convenient wireless setup and download via Bluetooth Low Energy (100-foot range)
  • Record a maximum 84650 measurments
  • Visual alarms alert you to out-of-range conditions
  • Compact, weatherproof housing with built-in mounting
  • View current data in real time with Onset’s free HOBOmobile app
  • Share downloaded data easily from your mobile device
  • Accuracy: ±0.2 °C
Leveraging Bluetooth Low Energy, the logger enables easy, fast set up and data download directly from your mobile device, and provides high-accuracy measurements in harsh outdoor environments.