HOBO® Bluetooth Low Energy Carbon Dioxide/Temp/RH Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
MX1102A
470323-468EA 672.21 USD
470323-468
HOBO® Bluetooth Low Energy Carbon Dioxide/Temp/RH Data Logger
Data Loggers
Onset’s HOBO MX1102 CO₂ logger makes it more convenient than ever to measure and record CO₂ in buildings and other non-condensing environments.
  • Wireless communication via Bluetooth Low Energy (BLE) Technology
  • Six month battery life at 5-minute CO₂ logging rate
  • Easy to deploy and offload using free HOBOmobile App
  • Visual and audible high & low alarm thresholds
  • Self-calibrating NDIR CO₂ sensor technology ensures optimal accuracy and lower maintenance costs
It measures CO₂ from 0 to 5000 parts per million (ppm) – and the free HOBOmobile app allows you to access data right from your mobile phone or tablet within a 100-foot range.