HOBO® Bluetooth Low Energy Temperature/Relative Humidity Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
MX1101
470323-466EA 176.79 USD
470323-466
HOBO® Bluetooth Low Energy Temperature/Relative Humidity Data Logger
Data Loggers
Onset’s HOBO MX1101 data logger measures and transmits temperature and relative humidity data wirelessly to mobile devices via Bluetooth Low Energy (BLE) technology.
  • Wireless communication via Bluetooth Low Energy (BLE) technology
  • Easy to deploy and offload using free HOBOmobile App
  • Visual and audible high & low alarm thresholds
  • Stores 84000 measurements
  • Accuracy: ±0.2 °C and ± 2% RH
  • Find me/pager feature
  • Aligned with GLOBE protocols
The self-contained wireless data logger, which works with Onset’s free HOBOmobile® app for logger setup and data management, enables you to access data anytime from mobile device for iOS or Android over a 100-foot range, and it requires no dedicated equipment beyond a mobile device for configuring the logger or reading out data.
Using Onset’s free HOBOmobile® app, you can view data in graphs, check the operational status of loggers, configure alarm notifications, and share data files.