HOBO® Water Level 30' Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
U20L-01
470323-410EA 436.48 USD
470323-410
HOBO® Water Level 30' Data Logger
Data Loggers
The HOBO® U20L-01 is a low-cost, research-grade water level data logger for continuously measuring water level and temperature in a wide range of underwater environments.
  • Self-contained non-vented design enables easy deployment
  • Ideal for use in both fresh and saltwater environments, including wells, streams, lakes, wetlands, and tidal areas
  • Depths up to 30 feet
  • Durable ceramic pressure sensor withstands freezing
  • HOBOware® Pro software provides easy conversion to accurate water level reading, fully compensated for barometric pressure, temperature, and water density
It features 0.1% measurement accuracy, a polypropylene housing for use in both fresh and salt water, and a non-vented design for convenient and hassle-free deployment.
Ordering information: This logger requires HOBOware® Pro software and either a U-DTW-1 Waterproof Shuttle or the Base-U-4 Optic USB Base Station for configuration and data offload. For more information on required HOBOware software (not included), contact your Avantor representative.