HOBO TidbiT® MX Temperature 5000' Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
MX2204
470323-388EA 183.39 USD
470323-388
HOBO TidbiT® MX Temperature 5000' Data Logger
Data Loggers
The HOBO MX2204 TidbiT waterproof temperature logger leverages the power of Bluetooth Low Energy (BLE) to deliver high-accuracy temperature measurements straight to your iOS or Android mobile device.
  • Convenient wireless setup and download via Bluetooth Low Energy (BLE)
  • Large memory stores 96000 measurements
  • Waterproof to 1500 meters (5000 feet)
  • Waterproof boot protects the logger during deployment
  • LED alarm notifies you when temperature exceeds set threshold
  • Water detection feature conserves battery life
  • Works with Onset’s free HOBOmobile app
Designed for durability, this rugged waterproof logger is ideal for long-term deployments in streams, lakes, and oceans. Data collection in the field has never been easier!