HOBO Water Level Sensor
Supplier: ONSET COMPUTER CORPORATION SE
MX2001-03-SS-S
470323-370EA 493.15 USD
470323-370
HOBO Water Level Sensor
Weather Stations
Measure water pressure, temperature, and water level measurement.
  • 3-point NIST traceable calibration certificate
  • Research level data brought to your device in real time
  • Burst pressure: 60 ft. depth
  • Pressure response time: <1 second
  • Size: 1" diameter, 3.9" length
These sensors are available in four depth ranges, and come in stainless-steel or titanium (recommended for use in saltwater) models.
The non-vented design reduces maintenance, and the durable ceramic sensor can withstand freezing.
Ordering information: Only functional with specific HOBO systems. Contact the Science Help Desk for additional information (sciencehelp@avantorsciences.com).