HOBO® Water Level Sensor
Supplier: ONSET COMPUTER CORPORATION SE
MX2001-02-SS-S
470323-366EA 442.89 USD
470323-366
HOBO® Water Level Sensor
Electrodes Other Electrodes
Research grade level measurements and data monitoring for water pressure, temperature, and water level measurements.
  • Non-vented design reduces maintenance
  • Durable, ceramic sensor
  • 3-point NIST-traceable calibration certificate
  • Maximum error: 0.1% FS, 1.0 cm (0.03') water
These sensors are available in four depth ranges, and come in stainless-steel or titanium (recommended for use in saltwater) models.
Ordering information: For use with HOBO MX2001 loggers, MicroRX Water Level Stations, RX3000 stations, and the HOBOnet Remote Water Level Monitoring System.