Backyard Brains® DIY Muscle Spikerbox
Supplier: BACKYARD BRAINS, INC.
BYBP0034
470323-210KT 102.65 USD
470323-210
Backyard Brains® DIY Muscle Spikerbox
Educational Materials Biology Educational Materials Physiology Learning Activities
Great student project.
  • Shipped as individual components for your students to assemble
  • Estimated time from 1.5 (hobbyist) to 4 (novice) hours
The muscle spiker box is now available in kit form to record EMGs. The kit takes ~4 hours (complete novice) to 1.5 hours (electronics hobbyist) to build. To build the kit, you will need a soldering iron, some solder, and wire cutters. A small magnifying glass to read the labels on the chips may be helpful for those of you with some mileage.