Rake Retractor
Supplier: United Scientific Supplies
RRET06
470321-010EA 11.15 USD
470321-010
Rake Retractor
Retractors
Easily manipulate heavier tissue.
  • Wide handle For easy manipulation
  • Three teeth provide a secure grip
  • Long handle provides space for students working in teams
Used for tissue manipulation during dissection, the rake retractor has three recurved teeth to pull tissue away from structures. Use of the retractor by an assistant, allows a student to make very clean and precise cuts separating the tissue from an underlying structure.