Clear Plastic Overflow Can
Supplier: United Scientific Supplies
OFLCN2
470320-742EA 9.5 USD
470320-742
Clear Plastic Overflow Can
Educational Materials Physics Educational Materials Mechanics Learning Activities
This transparent overflow can is graduated to 125 ml and includes an attached spout.
  • Clear design
Made of clear acrylic, the can measures 5.5 cm I.D.×7 cm high with a wall thickness of 3 mm.