Mini Projectile Launcher
Supplier: United Scientific Supplies
MPL001
470320-736EA 137.95 USD
470320-736
Mini Projectile Launcher
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Investigate horizontal and vertical motion.
  • Projectile range of two meters
  • Multiple launch velocities
This simple but precise launcher projects 16 mm steel balls at ranges suitable for use on the bench top or from the bench to the floor. A protractor and plumb line make it easy to set any required launch angle from 45° downwards to straight up.
Three different spring tensions give three repeatable launch speeds (ranges of 1 m, 1.5 m and 2 m) and a magnetic piston ensures that the ball does not touch the side of the barrel and launches straight every time.
A cable release gives a smooth triggering to prevent jerking. The unit can stand on its own or be fixed to the bench with a clamp.