Ward's® Incubating Shakers
Supplier: Ward's
H3501-E_WARD H3501_WARD H1001-M-E_WARD H1001-M_WARD H2012-E_WARD H2012_WARD H2010-E_WARD H2010_WARD
470320-570EA 12478.59 USD
470320-570 470320-572 470320-574 470320-576 470320-578 470320-580 470320-582 470320-584
Ward's® Incubating Shakers
Shakers and Mixers Shaking Incubators
These incubating shakers provide digitally regulated temperature with precise orbital mixing.
  • Great for cell culture or bacterial cell growth
  • Non-slip rubber platform mat included
  • Horizontally circular orbit for aeration and mixing
  • Optional platform available for use with MAGic Clamp™ system
  • Two year warranty
The XL Incu-Shaker is available with or without refrigeration. Mini Incu-Shaker is available with or without CO₂ incubation.
All units are compatible with the optional MAGic Clamp System which allows use of the MAGic Clamp™ attachment line, for quick-changes between flask clamps of different sizes. Additional purchase of MAGic Clamp Platform required. See product resource tab for list of compatible accessories.
Ordering information: Supplied with a non-slip, rubber coated surface, ideal for tissue culture flasks, petri dishes and staining trays. Optional platform available for use with MAGic Clamp™ system, this must be ordered separately.