Ward's® High Speed Microcentrifuges, 230 V
Supplier: MOLECULAR ARTS CORP SE
C2417-E_WARD
470320-562EA 2542.2 USD
470320-562
Ward's® High Speed Microcentrifuges, 230 V
Centrifuges Microcentrifuges
Touch screen, high speed centrifugation.
  • 230 V model
  • Full-color touch screen
  • One-button program recall
  • Combination rotor accepts tubes and strips
  • Compact design
  • Two-year warranty
Ward's High speed centrifuge features a next-generation control panel with touch screen. It keeps samples cool, even during extended runs at the maximum speed setting by a proprietary air flow system that moves air evenly and quietly throughout the chamber.
The unique combination-rotor accepts both standard (1.5 or 2.0 ml) microtubes and PCR tubes/strips, making this the perfect partner for any molecular biology laboratory.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (470320-564) is also available, please see separate product page.