Amino Labs DNA Extraction Kit
Supplier: AMINO LABS NORTH INCORPORATED SE
WWG001-S WWG001-G
470320-346EA 16.46 USD
470320-346 470320-348
Amino Labs DNA Extraction Kit
Educational Materials Biology Educational Materials Genetics Learning Activities
Visualize the DNA you eat every day.
  • Allows students to extract and visualize DNA from fruit
  • Quick and easy procedure takes only 15 minutes
  • Great beginner biotechnology activity
The incredible DNA extraction kit takes you through a journey of extracting the genomic DNA from the cells that make up a strawberry.
Additional items required but not included: 9 ml Isopropyl alcohol (99%), strawberry, plastic or glass cup).
Ordering information: Due to popular demand, we recommend you place your order at least a week in advance.
Delivery information: Though kits will ship at ambient temperature, some components will require refrigeration upon arrival. Refer to instructions.