Dlab Scientific Tube Rotators, Linear/Rotisserie Style
Supplier: DLAB SCIENTIFIC INC.
8031402000
470319-934EA 357.27 USD
470319-934
Dlab Scientific Tube Rotators, Linear/Rotisserie Style
Shakers and Mixers Rotating/Rocking Shakers
These rotators provide gentle and effective mixing of biological samples.
  • Units provide continuous operation
  • Supplied with a variety of attachments to hold tubes of different sizes
Time and speed display are represented in scale values.
Delivery information: Accessories 470319-936, 470319-938, 470319-940 are included with this product.