Stirring Hotplate
Supplier: DLAB SCIENTIFIC INC.
8030101212
470319-902EA 312.4 USD
470319-902
Stirring Hotplate
Stirrers Magnetic Hotplate Stirrers
An economically-priced LED hot plate stirrer, ideal to warm water to sub-boiling temperatures.
  • Digital temperature and speed control
  • Stainless steel heating plate with ceramic coating
  • Accommodates external temperature control
This compact unit is ideal to warm small volumes of water to near boiling temperatures (~90 °C) in an open beaker. To boil water in an open beaker, we suggest selecting one of our advanced hot plate stirrers with higher surface temperatures.
Accessories information: External temperature control probe and support rod are ordered separately.