Mini Block Heaters
Supplier: DLAB SCIENTIFIC INC.
18900415 18900416 18900428 5042101111 5042102111 5042132111 18900414 18900426 18900427
470319-524EA 158.58 USD
470319-524 470319-526 470319-528 470319-872 470319-874 470319-876 470319-878 470319-898 470319-900
Mini Block Heaters
Block Heaters Digital Block Heaters
Affordable mini dry-block heaters are a great option for the lab.
  • LCD Display of both temperature and time
  • Overheating protection
  • Rapid, 3-point self-calibration
  • USB Interface to save data
  • Available with or without lid/cooling
These single block mini heaters come with a block that can hold fifteen, 1.5 ml tubes, but with various additional blocks available, most common tubes can be used in these units. Three different varieties allow to choose whether or not you want to utilize a heated lid, and whether you want heating or heating and cooling. Choose the machine that best suits your needs.