High Speed Mini Centrifuge
Supplier: DLAB SCIENTIFIC INC.
9012002121
470319-480EA 1164.9 USD
470319-480
High Speed Mini Centrifuge
Centrifuges Microcentrifuges
The micro centrifuge is ideal for various application with its powerful centrifugal force.
  • Powerful centrifugal force of 15100×g
  • Brushless DC motor drive quickly accelerates the rotor to set speed
Built with safety in mind with safety devices like door interlock system to prevent the running during open, over speed detection, over temperature detection to prevent the rotor damage.
Having advanced features like automatic internal diagnosis, speed/RCF switch, short time run function and having sound alert function to alert the user.
Certifications: EMC, CE, FCC, ICES, cTUVus, CB certified. Accredited with CE, cTUVus, FCC certificates, and MCA test. MCA test according to IEC/EN 61010-2-20.
Delivery information: Includes A12-2P plastic rotor kit.