Microcentrifuges, 230 V, MC-24 Touch
Supplier: Ward's
C2417-E
470319-046EA 2743.8 USD
470319-046
Microcentrifuges, 230 V, MC-24 Touch
Centrifuges Microcentrifuges
Touch screen, high speed centrifugation.
  • 230 V model
  • Full-color touch screen
  • One-button program recall
  • Combination rotor accepts tubes and strips
  • Compact design
  • Two-year warranty
Benchmark’s MC-24 Touch is the first microcentrifuge in its class to feature touch screen control.
In addition to the next generation control panel, the centrifuge also offers high speed and a unique COMBI-Rotor™ that accepts both standard microtubes and PCR tubes/strips, making this the perfect partner for any molecular biology laboratory.
Caution: 230 V model.