Bone Clones® Sivapithecus Skull
Supplier: BONE CLONES HOLDINGS
BH-024
470317-794EA 304.6 USD
470317-794
Bone Clones® Sivapithecus Skull
Models Human Biology Models
8.5 to 12.5 MYA. The Sivapithecus indicus skull was discovered in 1979 by D. Pilbeam and S.M. Ibrahim Shah on the Potwar Plateau, Pakistan.
  • Each Bone Clones® cast is hand-made from start to finish
  • Rigorous standards are applied when selecting original specimens to ensure the best representation
  • Proprietary, custom blend of polyurethane developed to capture and replicate minute detail
This specimen consists of a nearly complete mandible (with complete dentition) and the left side of the face. The facial characteristics of Sivapithecus suggest a very orangutan-like appearance; however, characteristics in the dentition, such as thick enamel on the molars, point to similarities to early hominids. Recent study of postcranial remains indicates that Sivapithecus was not entirely orang-like. In recreating this important Late Miocene specimen, every effort was made to preserve all details, colors, and accurate measurements.
The Bone Clones® Hominid line is composed of discoveries from anatomically modern humans, archaic humans, early Homo, early hominins, and other hominids. The majority of the casts in this line have been recreated by a team of anatomical sculptors. Some are reconstructions made by anthropology professionals using fragmentary elements from original discoveries and extrapolating the missing parts from those.
Accessories information: Accessories not included.