Carson MicroFlip™ Pocket Microscope
Supplier: CARSON OPTICAL
MP-250
470317-026EA 30.3 USD
470317-026
Carson MicroFlip™ Pocket Microscope
Magnifiers
The MicroFlip™ MP-250 is an extremely powerful LED lighted pocket microscope that contains a unique flip-down slide base with an aspheric lens system for ultra precise image quality. The high power magnification ranges from 100x to 250x power. It also contains a built-in UV flashlight that is perfect for detecting counterfeit currency, nectar mapping and so much more.
Use the smartphone digiscoping clip to take pictures and record video of your subject matter using the built-in camera application on your smartphone. Then, share your images and videos on social media or email/text them to family, co-workers and friends.
The smartphone digiscoping clip will work with most smartphone models in the marketplace including iPhone, Samsung, HTC, LG and more. Thicker cases like Otterbox style cases may need to be removed before attaching the digiscoping clip. It is a great, cost effective educational field microscope for multiple hobbies, tasks, learning and exploration.
Fits all phones. For specific information on your model, visit: www.carson.com/multicam.
Ordering information: It is powered by 1 AA battery which is not included and it also comes included with a smartphone digiscoping clip, one starter slide with cover slip and a wrist strap for convenient portability.