Baseline® Windmill Spirometers
Supplier: Fabrication Enterprises
12-1710 12-1711 12-1714
470316-648CS 2141.3 USD
470316-648 470316-650 470316-652
Baseline® Windmill Spirometers
Respirators Respiratory Systems
Compact and lightweight.
  • Replacement mouthpieces in plastic or paper
Compact, lightweight spirometer for screening large populations. Subject blows into disposable mouthpiece. Indicator registers forced vital lung capacity up to 7 liters.
Delivery information: Includes 50 mouthpieces.