Neuro-Aide® Disposable Pinwheel
Supplier: Fabrication Enterprises
12-1455-25
470316-646PK 60.5 USD
470316-646
Neuro-Aide® Disposable Pinwheel
Clinical Diagnostic Supplies Clinical Instruments
Use a new wheel every time.
  • Disposable
  • Made of plastic
  • Compact size
Neuro-Aide neurological pinwheel consists of a rotating pinwheel attached to a handle. Used by orthopedists, neurologists, neurosurgeons, and other physicians to test a patient's ability to feel sharp contact with the skin. One can test for nerve damage, or nerve dysfunction.