Disposable Neurological Pinwheel
Supplier: Fabrication Enterprises
12-1451-25
470316-644PK 91.55 USD
470316-644
Disposable Neurological Pinwheel
Clinical Diagnostic Supplies Clinical Instruments
Use a new wheel every time.
  • Disposable sterile unit made of plastic
Use the pinwheel to elicit cutaneous sensory and pain perception responses.
Packaging: Individually packed.