Detecto® Bariatric/Wheelchair Scale
Supplier: Fabrication Enterprises
12-1355
470315-960EA 5693 USD
470315-960
Detecto® Bariatric/Wheelchair Scale
Balances and Scales
Easily weigh patients.
  • Easily position with built in handles
  • Pushbutton tare
  • BMI calculation
Accommodating up to 101 cm (40") wide wheelchairs, this American-made scale provides everything required for weighing bariatric patients. For added comfort, a patient may be wheeled onto the scale and the chair tare weight keyed in for an easy weighing. Heavy-duty, extra-large platform measuring 101×76 cm(40×30") with integral ramp, and 1,000 lbs. capacity makes this scale perfect for patients in manual or power wheelchairs, dialysis patients, or walk-ons. EMR/EHR ready with serial, USB, and optional ethernet connectivity.