ADC® Calibration Test Kit
Supplier: AMERICAN DIAGNOSTIC CORP CP
991
470315-738CS 272.6 USD
470315-738
ADC® Calibration Test Kit
Blood Pressure Monitors
Make sure your gauges are calibrated.
  • Not made of natural rubber latex
Do you think you have a manometer that needs calibration? Easily check to see if two devices agree using this simple calibration test kit. Simply connect a known good device, and a device in question, to the Y tubing, and apply pressure using the bulb. Both devices should give the same reading. If they don't you definitely need recalibration.
Delivery information: Includes connectors to permit testing of aneroid and mercury sphygs.