ADC® Prosphyg 775 Pocket Aneroid Sphygmomanometer
Supplier: AMERICAN DIAGNOSTIC CORP CP
775-11AN
470315-710CS 774.15 USD
470315-710
ADC® Prosphyg 775 Pocket Aneroid Sphygmomanometer
Blood Pressure Monitors
Perfect for institutional use.
  • Manometer exceeds industry standards
  • Cuff with range markings for accurate cuff selection
  • Not made with natural rubber latex
ADC®'s prosphyg 775 Series is an economical aneroid sphygmomanometer for today’s cost-conscious healthcare market. The black enamel 300 mm Hg manometer exceeds speciation's for accuracy and is backed by a ten year warranty. The navy blue nylon cuff features range markings to facilitate correct selection.
Delivery information: Leatherette case included.