ADC® Prosphyg 768 Pocket Aneroid Sphygmomanometer
Supplier: AMERICAN DIAGNOSTIC CORP CP
768-11ABK
470315-708CS 545.95 USD
470315-708
ADC® Prosphyg 768 Pocket Aneroid Sphygmomanometer
Blood Pressure Monitors
Affordable and professional.
  • Zippered color matched case for storage
  • Not made with natural rubber latex
ADC®'s prosphyg 768 series is our most popular sphygmomanometer. From the contemporary black enamel manometer to the meticulously designed adcuff inflation system, the 768 is the model of choice for professionals looking for high quality at an affordable price.
Delivery information: Includes 300 MMHG manometer, extra-large, color-matched, dual-pocket nylon carry case.