ADC® Advantage 6021N Automatic Digital Blood Pressure Monitor
Supplier: AMERICAN DIAGNOSTIC CORP CP
6021NSA
470315-676CS 1023.55 USD
470315-676
ADC® Advantage 6021N Automatic Digital Blood Pressure Monitor
Blood Pressure Monitors
Fully automatic readings.
  • Recalls readings for two users
  • Determines ideal inflation levels
  • Includes 5 year warranty
  • Not made with natural rubber latex
Digital monitors are a simple way for anyone to keep track of the blood pressure. This is an inexpensive, and easy to use solution. The cuff is marked for sizing and placement, and includes a d-ring allowing for one handed application. A single connection between the cuff and the display is all you need. The digital control unit controls inflation and deflation cycles, utilizing Smart Logic software to determine the ideal inflation level. The display shows systolic and diastolic pressures, as well as pulse and a warning if an irregular heart beat is detected. A memory can be accessed showing up to the last 99 readings, for up to two users.