Ward's® Essentials Standard Compound Microscope
Supplier: VWR
SFC100FLED
470315-506EA 163.45 USD
470315-506
Ward's® Essentials Standard Compound Microscope
Microscopes
Ideal for biological use and educational applications.
  • 40×, 100× and 400× magnification
  • Rechargeable LED illumination
  • Inclined rotatable head
  • 1 year limited warranty
This microscope model includes the most commonly specified features for standard high school instruction. The glass-coated DIN optics provide clear, crisp images and the rechargeable LED illumination offers added versatility and portability.