Ward's® Essentials Student Compound Microscope
Supplier: VWR
SW200
470315-504EA 99.95 USD
470315-504
Ward's® Essentials Student Compound Microscope
Microscopes
Quality performance at an economical price.
  • Fully rotatable head
  • 40 to 1000× with 4×, 10× and 40× obectives and 10× and 25× eyepieces
  • Adjustable LED illumination
  • All metal arm and base
  • One year limited warranty
Designed for elementary and middle school students, this microscope boasts the same features as many advanced models, but in a 3/4 size. The rugged design with metal arm and base, carry handle and cordless capability make this microscope a practical pick for classroom and field experiments.