Microdissection Scissors
Supplier: DR INSTRUMENTS INC. SE
9MCD
470315-228BX 309.95 USD
470315-228
Microdissection Scissors
Scissors
For extremely delicate dissection.
  • Spring form handle
For microdissection, these scissors work with a very light touch. The spring form handle keeps the blades open, and they operate like you are using forceps. The short blades allow for excellent control for the tightest locations.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.