Student Dissection Scissors
Supplier: DR INSTRUMENTS INC. SE
8
470315-162BX 97.9 USD
470315-162
Student Dissection Scissors
Scissors
Student grade.
  • Stainless steel construction
  • Sharp /sharp point configuration
Easier for students to use, these scissors are a durable stainless steel construction. Unlike a surgical style scissor, these have a gap between the handles when closed giving a shorter travel length for students cutting closer to the pivot point.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.