Vernier Go Direct® Charge Station
Supplier: Vernier Software & Technology
GDX-CRG
470315-120EA 88.99 USD
470315-120
Vernier Go Direct® Charge Station
Power Supplies
The Go Direct® charge station is the perfect solution for charging Go Direct sensors.
Each charge station has sixteen charging ports - eight USB and eight wand-style sensor ports. LED lights on the Go Direct sensors will show charging status. The Go Direct Charge Station is also compatible with Go Wireless devices.
Ordering information: USB cables not included.