BA310 Elite Upright Compound Microscopes
Supplier: MOTIC INSTRUMENTS USA, INC SE
1100100402645
470314-928EA 3482 USD
470314-928
BA310 Elite Upright Compound Microscopes
Microscopes
Built for the performance demands of colleges, universities and medical school markets.
  • Infinity corrected optics
  • CCIS EC plan achromatic objectives
  • Abbe condenser with iris diaphragm and slot (NA= 0.9/1.25)
The BA310 Elite model introduces superior improvements in both optical and mechanical performance. The rackless stage concept without a prominent gear rack permits even more convenient x/y movement and effortless positioning of the stage without gear rack interference. Ergonomic drop-down coaxial controls enable convenient 'eyes down' operation for uninterrupted sample viewing. The included CCIS EC Plan Achromatic lenses are multilayer-coated for improved color fidelity and image enhancement, giving superior image reproduction.
The improved BA310 Elite series introduces innovative features like second-generation EC Plan optics, a new rackless stage movement, and an advanced modular illumination design offering halogen/LED interchangeability. The BA310 Elite series brings versatility and function to a wide range of education and professional applications.
Ordering information: Microscope is supplied with all accessories listed above. Contact your Ward's representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.