Camera Adapters
Supplier: Motic
1101001901771 1101001901791 1101001901781 1101001904111 1101001901931
470314-900EA 199 USD
470314-900 470314-902 470314-904 470314-906 470314-908
Camera Adapters
Adapters Adapters, Camera to Microscope
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.