Horizon Energy Box
Supplier: HORIZON FUEL CELL TECHNOLOGIES
FCJJ-40
470314-476EA 1632.1 USD
470314-476
Horizon Energy Box
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Countless experiments, so many scientific principles at work and plenty of space for creativity.
  • Fuel cell science from fuel cell experts: PEM, direct ethanol, salt water and reversible fuel cells in one kit
  • Introduction to renewable energy: solar panel, wind turbine, temperature cell and hand crank
  • Includes super capacitor to demonstrate the latest in energy storage technology
  • Includes CD with curriculum content for 40 hours of classroom activities
The Renewable energy box provides a complete understanding of how fuel cell technology interacts with renewable energy sources to create an entirely sustainable power grid. Solar power, wind energy, kinetic energy from a hand crank and a demonstration of the incredible storage potential of a super capacitor. There’s a range of fuel cells to compare: PEM hydrogen fuel cell, the salt water fuel cell and a direct ethanol fuel cell.