Horizon Educational Vertical Axis Wind Turbine Science Kit
Supplier: HORIZON FUEL CELL TECHNOLOGIES
RESK-01
470314-472EA 135.19 USD
470314-472
Horizon Educational Vertical Axis Wind Turbine Science Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Teach renewable energy with this simple wind turbine design providing three different blade position combinations.
  • More than 25 hours of activities covering physics and earth/environmental science
  • Three different blade position combinations
  • Discover cutting-edge wind turbine technology
  • Students can also design their own blades
  • Hands on lab activities, teacher's guide, student's guides included
Delivery information: Included components: Three adjustable positions for the blades. Rotor base and mounting magnets. Axis assembly with removable coils. LED for output power demonstration, DIY (Do it yourself) blade set.