Horizon Wind to Hydrogen Science Kit
Supplier: HORIZON FUEL CELL TECHNOLOGIES
FCJJ-56
470314-468EA 394.51 USD
470314-468
Horizon Wind to Hydrogen Science Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Demonstrate the principles of wind energy, hydrogen fuel cells, electrolysis and basic electronics.
  • Turn wind power into hydrogen energy
  • Build your own hydrogen fuel cell powered inventions
  • Features revolutionary new Horizon wind turbine
  • Includes Horizon’s complete curriculum on renewable energy on CD
The Wind to Hydrogen Science Education Kit enables students to invent their own clean energy applications using a small electric motor powered by a fuel cell. Hydrogen is produced through water electrolysis by a reversible PEM fuel cell that is itself powered by a wind turbine with profiled blades based on NASA aeronautics. This hydrogen is then converted to electrical energy by the fuel cell and can be used to power a small fan.