Soil Formation Collection
Supplier: Avantor
470314-422KT 82.19 USD
470314-422
Soil Formation Collection
Educational Materials Earth Science Educational Materials Geology Learning Activities
Illustrate how soils are formed from parent rocks and minerals.
  • Includes 24 numbered specimens 1×1 to 1×2"
  • 8 Rock forming minerals, 8 rocks from which soils are derived, 3 examples of mechanical agents, 2 examples of chemical agents, and 4 soil types
Recommended for all courses where soil formation and composition are studied, this set includes samples of the various rocks and minerals from which solid is derived, as well as the soils themselves. The set is housed in a compartmented collection box, and specimen list describes the specimens and their role in soil formation.